Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277971.1 | NP_001264900.1 | 4 | 821 | ||
NM_032237.4 | NP_115613.1 | 5 | 917 | |||
GenBank mRNA | BC101548.1 | - | 3 | 688 | 8069054 | |
BC113703.1 | - | 3 | 688 | 8322755 |