Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032259.3 | NP_115635.1 | 3 | 2074 | ||
XM_011522699.2 | XP_011521001.1 | 4 | 2578 | |||
GenBank mRNA | AK301146.1 | - | 3 | 2058 | ||
AL136863.1 | - | 3 | 2051 | |||
BC008025.1 | - | 3 | 1539 | 3544791 | ||
BC009761.2 | - | 3 | 1074 |