Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032290.3 | NP_115666.2 | 21 | 2856 | ||
XM_017009978.1 | XP_016865467.1 | 21 | 2965 | |||
XM_017009979.1 | XP_016865468.1 | 21 | 7618 | |||
XM_017009980.1 | XP_016865469.1 | 20 | 2888 | |||
XM_017009981.1 | XP_016865470.1 | 21 | 2816 | |||
XM_017009983.1 | XP_016865472.1 | 11 | 1499 | |||
XM_017009984.1 | XP_016865473.1 | 15 | 1890 | |||
GenBank mRNA | AK299080.1 | - | 21 | 2882 | ||
AK309859.1 | - | 8 | 1052 | |||
AL136560.1 | - | 7 | 888 | |||
BC063674.1 | - | 7 | 1358 | 4368392 | ||
CR533539.1 | - | 7 | 868 | |||
CR936768.1 | - | 21 | 2862 |