Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033002.3 | NP_001028174.2 | 6 | 1084 | ||
NM_001160243.1 | NP_001153715.1 | 6 | 1084 | |||
NR_027679.1 | 5 | 972 | ||||
NR_027682.1 | 5 | 1020 | ||||
NR_027683.1 | 5 | 972 | ||||
NR_027685.1 | 4 | 908 | ||||
GenBank mRNA | AK096001.1 | - | 5 | 428 | ||
AK295394.1 | - | 6 | 525 | |||
AK300409.1 | - | 5 | 693 | |||
AY680654.1 | - | 6 | 514 | |||
AY680656.1 | - | 5 | 450 | |||
AY680657.1 | - | 5 | 402 | |||
AY680659.1 | - | 4 | 338 | |||
AY775314.1 | - | 6 | 1084 | |||
AY775315.1 | - | 5 | 1020 | |||
AY775317.1 | - | 5 | 1237 | |||
AY775320.1 | - | 4 | 908 | |||
AY775323.1 | - | 5 | 972 | |||
BC004451.1 | - | 5 | 463 | 2820882 | ||
BC013831.1 | - | 6 | 555 | |||
BC051849.2 | - | 6 | 517 | 5493799 | ||
DA349558.1 | - | 4 | 347 |