Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032314.3 | NP_115690.3 | 6 | 874 | ||
XM_006719639.2 | XP_006719702.1 | 7 | 974 | |||
GenBank mRNA | AK222610.1 | - | 6 | 864 | ||
AK225242.1 | - | 6 | 864 | |||
AK293656.1 | - | 6 | 895 | |||
AK298214.1 | - | 5 | 652 | |||
AK315681.1 | - | 6 | 865 | |||
BC004916.2 | - | 6 | 850 | |||
BC107874.1 | - | 6 | 868 | 6452052 | ||
BX647562.1 | - | 6 | 863 | |||
EU700459.1 | - | 6 | 868 |