Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NICN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032316.3 | NP_115692.1 | 1, 2 | 214 | ||
GenBank mRNA | AJ299740.1 | - | 1, 2 | 186 | ||
AK094248.1 | - | 1, 2 | 154 | |||
AK301846.1 | - | 1, 2 | 217 | |||
AK313073.1 | - | 1, 2 | 211 | |||
BC005050.1 | - | 1, 2 | 131 | 2820022 | ||
BC050653.1 | - | 1, 2 | 178 | 6015080 |