Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184992.1 | NP_001171921.1 | 1, 2 | 516 | ||
NM_032322.3 | NP_115698.3 | 1, 2 | 516 | |||
NM_197939.1 | NP_922921.1 | 1, 2 | 516 | |||
GenBank mRNA | AB470605.1 | - | 1, 2 | 380 | ||
AJ496729.1 | - | 1, 2 | 431 | |||
AK122646.1 | - | 1, 2 | 389 | |||
AK122709.1 | - | 2, 3 | 311 | |||
AK312979.1 | - | 1, 2 | 392 | |||
AY598332.1 | - | 1, 2 | 380 | |||
BC005084.1 | - | 1, 2 | 431 | |||
BC082262.1 | - | 1, 2 | 394 | |||
BC126420.1 | - | 1, 2 | 470 | 8992136 | ||
BC126422.1 | - | 1, 2 | 470 | 8992138 |