Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032323.2 | NP_115699.1 | 2 | 831 | ||
NR_026678.1 | 2 | 856 | ||||
GenBank mRNA | AK056816.1 | - | 2 | 796 | ||
AK122634.1 | - | 3 | 577 | |||
AK315767.1 | - | 2 | 799 | |||
BC005094.1 | - | 2 | 821 | 3955668 |