Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128612.2 | NP_001122084.1 | 2 | 192 | ||
NM_001128613.2 | NP_001122085.1 | 2 | 306 | |||
NM_001271831.1 | NP_001258760.1 | 2 | 192 | |||
NM_032344.3 | NP_115720.1 | 2 | 306 | |||
GenBank mRNA | AK308555.1 | - | 2 | 280 | ||
BC006129.1 | - | 2 | 290 | 3625545 | ||
BG705568.1 | - | 2 | 197 | |||
BM045237.1 | - | 2 | 292 | |||
BM923060.1 | - | 2 | 191 | |||
HY075773.1 | - | 2 | 176 | |||
HY160553.1 | - | 2 | 300 |