Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040160.2 | NP_001035250.1 | 3 | 454 | ||
NM_001040162.2 | NP_001035252.1 | 2 | 291 | |||
NM_032366.4 | NP_115742.3 | 3 | 454 | |||
NR_109976.1 | 2 | 291 | ||||
NR_109977.1 | 2 | 291 | ||||
NR_109978.1 | 3 | 454 | ||||
XM_011522713.2 | XP_011521015.1 | 3 | 546 | |||
XM_011522714.2 | XP_011521016.1 | 3 | 546 | |||
GenBank mRNA | AK057077.1 | - | 1 | 1028 | ||
AK293046.1 | - | 1 | 1014 | |||
AK294482.1 | - | 3 | 390 | |||
AK295628.1 | - | 3 | 390 | |||
AW027232.1 | - | 2 | 203 | |||
BG612714.1 | - | 2 | 256 | |||
BI762987.1 | - | 3 | 324 | |||
BM827705.1 | - | 2 | 226 | |||
BP276058.1 | - | 3 | 421 | |||
BQ921757.1 | - | 359 | ||||
BX104467.1 | - | 4 | 416 | |||
CK904040.1 | - | 3 | 358 |