Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040160.2 | NP_001035250.1 | 5 | 596 | ||
NM_001040161.2 | NP_001035251.1 | 3 | 373 | |||
NM_001040162.2 | NP_001035252.1 | 4 | 433 | |||
NM_001040165.2 | NP_001035255.1 | 4 | 536 | |||
NM_001288710.1 | NP_001275639.1 | 5 | 604 | |||
NM_032366.4 | NP_115742.3 | 6 | 664 | |||
NR_109976.1 | 5 | 608 | ||||
NR_109977.1 | 5 | 501 | ||||
NR_109978.1 | 6 | 771 | ||||
NR_109979.1 | 4 | 441 | ||||
XM_011522713.2 | XP_011521015.1 | 6 | 863 | |||
XM_011522714.2 | XP_011521016.1 | 5 | 688 | |||
GenBank mRNA | AI146806.1 | - | 4 | 430 | ||
AK057077.1 | - | 3 | 1562 | |||
AK294482.1 | - | 6 | 600 | |||
AK295628.1 | - | 5 | 532 | |||
BC007207.1 | - | 3 | 355 | 4054799 | ||
BI762987.1 | - | 6 | 640 | |||
BM827705.1 | - | 4 | 368 | |||
BP276058.1 | - | 5 | 563 | |||
BQ921757.1 | - | 569 | ||||
BU539308.1 | - | 4 | 389 | |||
BX104467.1 | - | 1 | 206 |