Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
siRNA ID
HSS189074
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
7
876
NM_001130084.1
NP_001123556.1
7
876
NM_001130085.1
NP_001123557.1
7
876
NM_001130086.1
NP_001123558.1
7
876
NM_001130087.1
NP_001123559.1
7
876
NM_001130088.1
NP_001123560.1
7
876
NM_001286688.1
NP_001273617.1
2
306
NM_032432.4
NP_115808.3
7
876
XM_005248014.4
XP_005248071.1
7
847
XM_005248017.4
XP_005248074.1
7
847
XM_005248018.4
XP_005248075.1
7
846
XM_005248019.4
XP_005248076.1
7
846
XM_005248020.4
XP_005248077.1
7
846
XM_005248021.4
XP_005248078.1
7
845
XM_005248022.4
XP_005248079.1
7
846
XM_005248023.4
XP_005248080.1
7
846
XM_005248024.4
XP_005248081.1
7
845
XM_005248027.4
XP_005248084.1
7
845
XM_005248028.4
XP_005248085.1
7
842
XM_005248029.4
XP_005248086.1
7
844
XM_005248030.4
XP_005248087.1
7
844
XM_005248031.4
XP_005248088.1
7
846
XM_006713923.3
XP_006713986.1
7
847
XM_006713924.3
XP_006713987.1
7
846
XM_006713925.3
XP_006713988.1
7
847
XM_006713927.3
XP_006713990.1
7
845
XM_011513586.2
XP_011511888.1
7
851
XM_017008706.1
XP_016864195.1
7
848
XM_017008707.1
XP_016864196.1
7
857
XM_017008708.1
XP_016864197.1
7
846
XM_017008709.1
XP_016864198.1
7
847
XM_017008710.1
XP_016864199.1
7
846
XM_017008711.1
XP_016864200.1
7
846
XM_017008712.1
XP_016864201.1
7
846
XM_017008713.1
XP_016864202.1
7
845
XM_017008714.1
XP_016864203.1
7
857
XM_017008715.1
XP_016864204.1
7
846
XM_017008716.1
XP_016864205.1
7
845
XM_017008717.1
XP_016864206.1
7
845
XM_017008718.1
XP_016864207.1
7
857
XM_017008719.1
XP_016864208.1
7
845
XM_017008720.1
XP_016864209.1
7
844
XM_017008721.1
XP_016864210.1
7
844
XM_017008722.1
XP_016864211.1
7
845
XM_017008723.1
XP_016864212.1
7
857
XM_017008724.1
XP_016864213.1
7
857
XM_017008725.1
XP_016864214.1
7
857
XM_017008726.1
XP_016864215.1
2
306
XM_017008727.1
XP_016864216.1
2
306
GenBank mRNA
AB058711.1
-
6
635
AJ748600.1
-
7
798
AJ748601.1
-
7
798
AK094754.1
-
7
855
AK094798.1
-
2
276
AK301060.1
-
7
839
BC067214.1
-
7
888
BC122567.1
-
7
827
40085570
DA392726.1
-
2
306
DQ413176.1
-
7
753
DQ413177.1
-
6
733
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0