Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032435.2 | NP_115811.2 | 9 | 2478 | ||
XM_011544305.2 | XP_011542607.1 | 9 | 2514 | |||
GenBank mRNA | AB058707.1 | - | 3 | 675 | ||
AJ311798.1 | - | 9 | 2478 | |||
AK309811.1 | - | 1 | 31 | |||
BC136648.1 | - | 9 | 2337 | 9020638 | ||
BC136649.1 | - | 9 | 2337 | 9020639 |