Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX6-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177400.2 | NP_796374.1 | 3 | 787 | ||
XM_017016789.1 | XP_016872278.1 | 3 | 781 | |||
GenBank mRNA | BC101635.1 | - | 3 | 787 | 8069141 | |
BC104823.1 | - | 3 | 787 | 8143826 | ||
BF509028.1 | - | 3 | 417 |