Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 18 | 2793 | ||
NM_001281521.1 | NP_001268450.1 | 18 | 2859 | |||
NM_001281522.1 | NP_001268451.1 | 17 | 2598 | |||
NM_032485.5 | NP_115874.3 | 18 | 2739 | |||
NM_182802.2 | NP_877954.1 | 18 | 2745 | |||
XM_017028105.1 | XP_016883594.1 | 18 | 2888 | |||
XM_017028106.1 | XP_016883595.1 | 18 | 2705 | |||
XM_017028107.1 | XP_016883596.1 | 11 | 2978 | |||
GenBank mRNA | AJ439063.1 | - | 18 | 2739 | ||
AK027644.1 | - | 12 | 1778 | |||
AK314654.1 | - | 18 | 2421 | |||
AY158211.1 | - | 18 | 2444 | |||
BC005170.1 | - | 18 | 2444 | |||
BC008830.2 | - | 15 | 2049 | |||
BC080656.1 | - | 18 | 2444 | 4841271 | ||
BC101055.1 | - | 18 | 2660 | 40008095 | ||
BC101056.1 | - | 18 | 2540 | 40008096 | ||
BC101057.1 | - | 18 | 2594 | 40008097 | ||
KU178803.1 | - | 12 | 1651 | |||
KU178804.1 | - | 13 | 1742 | |||
KU178805.1 | - | 11 | 1510 |