Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202406.1 | NP_001189335.1 | 6 | 936 | ||
NM_001202407.1 | NP_001189336.1 | 6 | 946 | |||
NM_001202408.1 | NP_001189337.1 | 6 | 1014 | |||
NM_001202409.1 | NP_001189338.1 | 7 | 1236 | |||
NM_001202410.1 | NP_001189339.1 | 7 | 1084 | |||
NM_001202411.1 | NP_001189340.1 | 7 | 1113 | |||
NM_001202412.1 | NP_001189341.1 | 7 | 799 | |||
NM_032497.2 | NP_115886.1 | 7 | 999 | |||
GenBank mRNA | AB075488.1 | - | 2 | 299 | ||
AK027700.1 | - | 7 | 677 | |||
AK056489.1 | - | 6 | 511 | |||
AK092348.1 | - | 5 | 1033 | |||
AK298169.1 | - | 4 | 290 | |||
AK308517.1 | - | 6 | 607 | |||
AK310020.1 | - | 7 | 811 | |||
AK310117.1 | - | 6 | 1102 | |||
AK310781.1 | - | 4 | 931 | |||
BC006436.1 | - | 6 | 984 | 3957973 | ||
DA509657.1 | - | 3 | 224 | |||
DA774704.1 | - | 5 | 413 |