Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130010.2 | NP_001123482.1 | 10 | 892 | ||
NM_001290232.1 | NP_001277161.1 | 10 | 583 | |||
NM_001290233.1 | NP_001277162.1 | 10 | 892 | |||
NM_001321756.1 | NP_001308685.1 | 9 | 846 | |||
NM_001321757.1 | NP_001308686.1 | 11 | 926 | |||
NM_001321758.1 | NP_001308687.1 | 8 | 781 | |||
NM_001321759.1 | NP_001308688.1 | 10 | 892 | |||
NM_001321761.1 | NP_001308690.1 | 10 | 910 | |||
NM_032499.5 | NP_115888.1 | 11 | 638 | |||
XM_011522111.1 | XP_011520413.1 | 10 | 884 | |||
XM_017022676.1 | XP_016878165.1 | 7 | 686 | |||
XM_017022677.1 | XP_016878166.1 | 7 | 686 | |||
GenBank mRNA | AF114263.1 | - | 11 | 613 | ||
AK026504.1 | - | 10 | 861 | |||
AK315446.1 | - | 10 | 552 | |||
BC006254.2 | - | 10 | 651 | 3952242 | ||
BC042750.1 | - | 2 | 4997 | |||
BX647650.1 | - | 4 | 262 |