Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129993.1 | NP_001123465.1 | 3 | 362 | ||
NM_032506.2 | NP_115895.2 | 3 | 436 | |||
XM_005264605.3 | XP_005264662.1 | 3 | 363 | |||
XM_005264606.4 | XP_005264663.1 | 3 | 363 | |||
XM_005264607.2 | XP_005264664.1 | 3 | 343 | |||
XM_011533126.2 | XP_011531428.1 | 3 | 415 | |||
XM_011533127.2 | XP_011531429.1 | 3 | 275 | |||
XM_011533128.1 | XP_011531430.1 | 3 | 363 | |||
XM_011533129.2 | XP_011531431.1 | 3 | 363 | |||
XM_011533131.2 | XP_011531433.1 | 3 | 363 | |||
XM_017005108.1 | XP_016860597.1 | 3 | 277 | |||
XM_017005109.1 | XP_016860598.1 | 3 | 363 | |||
GenBank mRNA | AK131267.1 | - | 4 | 409 | ||
AK309749.1 | - | 3 | 344 | |||
AL833595.1 | - | 3 | 301 | |||
BC039298.1 | - | 3 | 353 | 5314819 | ||
BC070104.1 | - | 3 | 436 | 5268265 | ||
BX647395.1 | - | 3 | 253 | |||
DA222969.1 | - | 3 | 362 | |||
DB086916.1 | - | 3 | 454 | |||
DB090584.1 | - | 3 | 416 |