Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195483.1 | NP_001182412.1 | 3 | 416 | ||
NM_024628.5 | NP_078904.3 | 4 | 477 | |||
GenBank mRNA | AF345197.1 | - | 3 | 366 | ||
AF390442.1 | - | 3 | 417 | |||
AK123001.1 | - | 4 | 431 | |||
BC126158.1 | - | 3 | 387 | 8991874 | ||
BC126160.1 | - | 3 | 387 | 8991876 | ||
DA858738.1 | - | 4 | 477 |