Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003594.3 | NP_003585.3 | 1, 2 | 52 | ||
XM_005271277.3 | XP_005271334.1 | 1, 2 | 91 | |||
XM_011542303.2 | XP_011540605.1 | 1, 2 | 91 | |||
XM_017002549.1 | XP_016858038.1 | 1, 2 | 91 | |||
XM_017002550.1 | XP_016858039.1 | 1, 2 | 91 | |||
XM_017002551.1 | XP_016858040.1 | 1, 2 | 91 | |||
XM_017002552.1 | XP_016858041.1 | 1, 2 | 91 | |||
XM_017002553.1 | XP_016858042.1 | 1, 2 | 91 | |||
XM_017002554.1 | XP_016858043.1 | 1, 2 | 91 | |||
XM_017002555.1 | XP_016858044.1 | 1, 2 | 91 | |||
GenBank mRNA | AF073771.1 | - | 1, 2 | 52 | ||
AF080255.1 | - | 1, 2 | 62 | |||
AK291017.1 | - | 1, 2 | 91 | |||
BC030058.1 | - | 1, 2 | 36 | 5457833 |