Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083113.1 | NP_001076582.1 | 2, 3 | 721 | ||
NM_001195653.1 | NP_001182582.1 | 2, 3 | 745 | |||
NM_001195654.1 | NP_001182583.1 | 2, 3 | 764 | |||
NM_032527.4 | NP_115916.3 | 2, 3 | 764 | |||
NM_181485.2 | NP_852150.2 | 2, 3 | 694 | |||
GenBank mRNA | AB058750.3 | - | 2, 3 | 698 | ||
AK027878.1 | - | 2, 3 | 702 | |||
AK314726.1 | - | 2, 3 | 734 | |||
BC019338.1 | - | 2, 3 | 739 | 2906985 | ||
BC032612.1 | - | 2, 3 | 684 | 5555076 | ||
CV027894.1 | - | 1, 2 | 575 | |||
KU178806.1 | - | 1, 2 | 575 | |||
KU178807.1 | - | 1, 2 | 575 |