Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1F10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032556.5 | NP_115945.4 | 2, 3 | 547 | ||
NM_173161.2 | NP_775184.1 | 3, 4 | 201 | |||
GenBank mRNA | AY029413.1 | - | 3, 4 | 201 | ||
AY358846.1 | - | 3, 4 | 189 |