Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RETNLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032579.2 | NP_115968.1 | 1, 2 | 215 | ||
GenBank mRNA | AF290873.1 | - | 1, 2 | 217 | ||
AF323084.1 | - | 1, 2 | 224 | |||
AM050721.1 | - | 1, 2 | 119 | |||
AY358655.1 | - | 1, 2 | 216 | |||
BC069318.1 | - | 1, 2 | 188 | 7262095 | ||
BC113502.1 | - | 1, 2 | 149 | 8322554 | ||
BC113528.1 | - | 1, 2 | 149 | 8322580 |