Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM126A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032581.3 | NP_115970.2 | 11 | 1749 | ||
XM_005249894.3 | XP_005249951.1 | 12 | 2089 | |||
XM_005249895.2 | XP_005249952.1 | 12 | 2008 | |||
XM_011515589.2 | XP_011513891.1 | 11 | 1600 | |||
GenBank mRNA | AB030241.1 | - | 11 | 1655 | ||
AK054887.1 | - | 2 | 532 | |||
AK056319.1 | - | 11 | 1686 | |||
BC018710.1 | - | 11 | 1727 | 4649785 | ||
BX640757.1 | - | 1 | 2047 | |||
KU178808.1 | - | 10 | 1494 | |||
KU178809.1 | - | 11 | 1790 | |||
KU178810.1 | - | 11 | 1709 |