Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM126A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032581.3 | NP_115970.2 | 5 | 648 | ||
XM_005249894.3 | XP_005249951.1 | 5 | 692 | |||
XM_005249895.2 | XP_005249952.1 | 5 | 692 | |||
XM_011515589.2 | XP_011513891.1 | 5 | 499 | |||
XM_011515590.1 | XP_011513892.1 | 5 | 692 | |||
GenBank mRNA | AB030241.1 | - | 5 | 554 | ||
AK056319.1 | - | 5 | 585 | |||
AK297399.1 | - | 4 | 483 | |||
AL833296.1 | - | 4 | 516 | |||
BC018710.1 | - | 5 | 626 | 4649785 | ||
KU178808.1 | - | 4 | 393 | |||
KU178809.1 | - | 4 | 393 | |||
KU178810.1 | - | 4 | 393 |