Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 1765 | ||
NM_001172675.1 | NP_001166146.1 | 5 | 1445 | |||
NM_032584.2 | NP_115973.2 | 5 | 1442 | |||
XM_005259335.4 | XP_005259392.1 | 5 | 1764 | |||
XM_005259336.4 | XP_005259393.1 | 5 | 1554 | |||
XM_017027384.1 | XP_016882873.1 | 5 | 1784 | |||
XM_017027385.1 | XP_016882874.1 | 5 | 1468 | |||
XM_017027386.1 | XP_016882875.1 | 4 | 1390 | |||
GenBank mRNA | AK095593.1 | - | 1 | 1003 | ||
AK096623.1 | - | 5 | 1767 | |||
AK289994.1 | - | 5 | 1429 | |||
AK314603.1 | - | 5 | 1426 | |||
AL713691.1 | - | 1 | 424 | |||
AY029765.1 | - | 5 | 1404 | |||
BC136255.1 | - | 4 | 1394 | 9020242 | ||
BC143412.1 | - | 4 | 1397 |