Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032587.3 | NP_115976.2 | 3 | 935 | ||
XM_017009989.1 | XP_016865478.1 | 2 | 488 | |||
XM_017009990.1 | XP_016865479.1 | 4 | 1169 | |||
GenBank mRNA | AB209844.1 | - | 3 | 755 | ||
AF356193.1 | - | 3 | 1059 | |||
AK304349.1 | - | 3 | 867 | |||
BC093825.1 | - | 3 | 882 | 7939670 |