Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032609.2 | NP_115998.2 | 5 | 570 | ||
XM_005260579.4 | XP_005260636.1 | 4 | 572 | |||
XM_005260580.4 | XP_005260637.1 | 3 | 453 | |||
XM_005260581.3 | XP_005260638.1 | 4 | 470 | |||
GenBank mRNA | AF257180.1 | - | 5 | 570 | ||
BC057779.1 | - | 5 | 546 | 30337282 | ||
HE647859.1 | - | 4 | 495 |