Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290149.1 | NP_001277078.1 | 6 | 1650 | ||
NM_001290150.1 | NP_001277079.1 | 3 | 1081 | |||
NM_001290151.1 | NP_001277080.1 | 4 | 1134 | |||
NM_001290152.1 | NP_001277081.1 | 4 | 1111 | |||
NM_001290153.1 | NP_001277082.1 | 5 | 1272 | |||
NM_001290154.1 | NP_001277083.1 | 5 | 1328 | |||
NM_001290155.1 | NP_001277084.1 | 4 | 1167 | |||
NM_032712.2 | NP_116101.1 | 4 | 951 | |||
NM_199249.2 | NP_954857.1 | 5 | 1306 | |||
NM_199250.2 | NP_954858.1 | 4 | 1145 | |||
GenBank mRNA | AY491972.1 | - | 1 | 769 | ||
BC006151.1 | - | 2 | 802 | 3354369 | ||
BC037227.1 | - | 3 | 868 | 3446932 | ||
BC051842.1 | - | 5 | 1350 | 5110472 |