Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032737.3 | NP_116126.3 | 10 | 1717 | ||
GenBank mRNA | BC006551.2 | - | 10 | 1682 | 2961134 | |
BT007441.1 | - | 10 | 1622 | |||
KP137589.1 | - | 7 | 1271 | |||
M94362.1 | - | 9 | 1367 |