Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032752.1 | NP_116141.1 | 4, 5 | 1045 | ||
XM_005273328.3 | XP_005273385.1 | 5, 6 | 2536 | |||
XM_005273330.3 | XP_005273387.1 | 5, 6 | 2536 | |||
XM_006711838.2 | XP_006711901.1 | 5, 6 | 2536 | |||
XM_011544310.1 | XP_011542612.1 | 5, 6 | 2536 | |||
XM_011544312.1 | XP_011542614.1 | 5, 6 | 2536 | |||
XM_011544314.2 | XP_011542616.1 | 5, 6 | 2536 | |||
XM_011544315.2 | XP_011542617.1 | 5, 6 | 2536 | |||
XM_011544316.1 | XP_011542618.1 | 5, 6 | 2536 | |||
XM_017002586.1 | XP_016858075.1 | 5, 6 | 2536 | |||
XM_017002587.1 | XP_016858076.1 | 5, 6 | 2489 | |||
XM_017002588.1 | XP_016858077.1 | 5, 6 | 2536 | |||
XM_017002589.1 | XP_016858078.1 | 5, 6 | 2536 | |||
XM_017002590.1 | XP_016858079.1 | 5, 6 | 2536 | |||
XM_017002591.1 | XP_016858080.1 | 5, 6 | 2489 | |||
XM_017002592.1 | XP_016858081.1 | 5, 6 | 2536 | |||
XM_017002593.1 | XP_016858082.1 | 5, 6 | 2536 | |||
GenBank mRNA | BC007263.1 | - | 4, 5 | 1045 | 3051320 | |
BC025794.1 | - | 2, 3 | 247 |