Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039970.1 | NP_001035059.1 | 11, 12 | 1542 | ||
NM_001278236.1 | NP_001265165.1 | 11, 12 | 2376 | |||
NM_001278238.1 | NP_001265167.1 | 10, 11 | 1505 | |||
NM_001278239.1 | NP_001265168.1 | 10, 11 | 1409 | |||
NM_006906.1 | NP_008837.1 | 11, 12 | 1638 | |||
NM_032781.3 | NP_116170.3 | 11, 12 | 1593 | |||
XM_011520411.2 | XP_011518713.1 | 11, 12 | 1577 | |||
XM_017018434.1 | XP_016873923.1 | 11, 12 | 1577 | |||
XM_017018435.1 | XP_016873924.1 | 11, 12 | 1547 | |||
XM_017018436.1 | XP_016873925.1 | 10, 11 | 1483 | |||
XM_017018437.1 | XP_016873926.1 | 11, 12 | 1501 | |||
XM_017018438.1 | XP_016873927.1 | 9, 10 | 1262 | |||
XM_017018439.1 | XP_016873928.1 | 10, 11 | 1388 | |||
XM_017018440.1 | XP_016873929.1 | 11, 12 | 1577 | |||
XM_017018441.1 | XP_016873930.1 | 11, 12 | 1577 | |||
GenBank mRNA | AK090923.1 | - | 11, 12 | 1502 | ||
AK127312.1 | - | 11, 12 | 2376 | |||
AK294374.1 | - | 10, 11 | 1245 | |||
AK294673.1 | - | 10, 11 | 1206 | |||
AK295604.1 | - | 10, 11 | 1366 | |||
AK316270.1 | - | 10, 11 | 1290 | |||
AL832541.1 | - | 11, 12 | 1638 | |||
BC039897.1 | - | 10, 11 | 1301 | |||
BC046435.1 | - | 1, 2 | 117 | |||
BC064807.1 | - | 11, 12 | 1402 | 5726473 |