Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGBL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323573.1 | NP_001310502.1 | 10 | 1285 | ||
NM_001323574.1 | NP_001310503.1 | 10 | 1285 | |||
NM_001323575.1 | NP_001310504.1 | 10 | 1249 | |||
NM_032785.3 | NP_116174.3 | 10 | 1249 | |||
NR_136623.1 | 10 | 1248 | ||||
XM_011542308.2 | XP_011540610.1 | 11 | 1344 | |||
XM_017002595.1 | XP_016858084.1 | 10 | 1306 | |||
XM_017002596.1 | XP_016858085.1 | 10 | 1306 | |||
XM_017002597.1 | XP_016858086.1 | 10 | 1306 | |||
GenBank mRNA | AK027348.1 | - | 6 | 628 | ||
AK094826.1 | - | 10 | 1208 | |||
AK294394.1 | - | 10 | 1244 | |||
AK294633.1 | - | 10 | 1248 | |||
AK296221.1 | - | 9 | 1065 | |||
BC126383.1 | - | 6 | 561 | 8992099 | ||
BC144171.1 | - | 5 | 471 |