Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009182.1 | NP_001009182.1 | 7 | 629 | ||
NM_001009183.1 | NP_001009183.1 | 8 | 674 | |||
NM_003616.2 | NP_003607.1 | 8 | 674 | |||
XM_017021709.1 | XP_016877198.1 | 7 | 652 | |||
GenBank mRNA | AB037701.1 | - | 7 | 607 | ||
AB037702.1 | - | 8 | 652 | |||
AB037703.1 | - | 8 | 572 | |||
AF027150.1 | - | 8 | 735 | |||
AK313947.1 | - | 8 | 662 | |||
BC028095.1 | - | 9 | 791 | |||
BC104968.1 | - | 8 | 673 | 8143971 |