Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNFT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109903.1 | NP_001103373.1 | 4 | 764 | ||
NM_032814.3 | NP_116203.2 | 4 | 764 | |||
GenBank mRNA | AK027533.1 | - | 5 | 800 | ||
BC011878.2 | - | 4 | 744 | 3532898 | ||
BQ676121.1 | - | 1 | 435 |