Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282933.1 | NP_001269862.1 | 3 | 436 | ||
NM_032819.4 | NP_116208.3 | 3 | 436 | |||
NR_104259.1 | 3 | 436 | ||||
GenBank mRNA | AK122943.1 | - | 3 | 539 | ||
BC132873.1 | - | 3 | 236 | 40146895 | ||
BC144188.1 | - | 3 | 236 | 9052708 | ||
BC146811.1 | - | 3 | 198 | 8860372 | ||
BC157823.1 | - | 3 | 236 | 9057042 | ||
DB091229.1 | - | 3 | 436 |