Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM222A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032829.2 | NP_116218.2 | 3 | 1905 | ||
XM_006719654.2 | XP_006719717.1 | 3 | 1491 | |||
XM_017020055.1 | XP_016875544.1 | 3 | 1875 | |||
GenBank mRNA | AK027627.1 | - | 3 | 1509 | ||
AK074806.1 | - | 1 | 1164 | |||
BC020193.1 | - | 3 | 1902 | |||
BC037221.1 | - | 2 | 1380 | 5197135 |