Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077268.1 | NP_001070736.1 | 2 | 828 | ||
NM_001258420.1 | NP_001245349.1 | 2 | 828 | |||
NM_001258421.1 | NP_001245350.1 | 2 | 631 | |||
NM_032850.4 | NP_116239.3 | 3 | 497 | |||
XM_017022683.1 | XP_016878172.1 | 1 | 1473 | |||
XM_017022684.1 | XP_016878173.1 | 2 | 815 | |||
GenBank mRNA | AB209224.1 | - | 2063 | |||
AF445414.1 | - | 3 | 339 | |||
AK027746.1 | - | 2 | 260 | |||
AK122779.1 | - | 2 | 618 | |||
AK225553.1 | - | 2 | 251 | |||
DB012963.1 | - | 2 | 246 |