Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032854.3 | NP_116243.2 | 4 | 466 | ||
XM_005258047.3 | XP_005258104.1 | 5 | 748 | |||
XM_005258048.4 | XP_005258105.2 | 5 | 685 | |||
XM_011525385.2 | XP_011523687.1 | 5 | 685 | |||
XM_011525386.2 | XP_011523688.1 | 5 | 685 | |||
XM_011525387.2 | XP_011523689.1 | 4 | 562 | |||
XM_011525388.2 | XP_011523690.1 | 5 | 685 | |||
XM_011525389.2 | XP_011523691.1 | 5 | 685 | |||
XM_017025235.1 | XP_016880724.1 | 4 | 629 | |||
GenBank mRNA | AK096431.1 | - | 4 | 371 | ||
AL833952.1 | - | 3 | 429 | |||
BI758263.1 | - | 5 | 579 |