Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032871.3 | NP_116260.2 | 6 | 728 | ||
NM_152222.1 | NP_689408.1 | 6 | 666 | |||
XM_011545305.1 | XP_011543607.1 | 5 | 551 | |||
GenBank mRNA | AF319553.1 | - | 6 | 728 | ||
AK027899.1 | - | 6 | 666 | |||
AK074128.1 | - | 1 | 363 | |||
AK310103.1 | - | 3 | 233 | |||
BC017279.2 | - | 6 | 591 | 4873951 | ||
BC051810.1 | - | 6 | 618 | 5216988 |