Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281504.1 | NP_001268433.1 | 6 | 1162 | ||
NM_032885.5 | NP_116274.3 | 6 | 1088 | |||
NR_104024.1 | 6 | 1044 | ||||
NR_104025.1 | 5 | 811 | ||||
XM_006722927.1 | XP_006722990.1 | 4 | 743 | |||
GenBank mRNA | AJ312332.1 | - | 6 | 904 | ||
AK056210.1 | - | 6 | 1063 | |||
AK294676.1 | - | 6 | 1162 | |||
AK314712.1 | - | 6 | 1023 | |||
BC007639.1 | - | 5 | 734 | |||
BC068992.1 | - | 6 | 1064 | 6578102 |