Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190726.1 | NP_001177655.1 | 2 | 419 | ||
NM_032918.2 | NP_116307.1 | 2 | 419 | |||
XM_017020121.1 | XP_016875610.1 | 2 | 745 | |||
GenBank mRNA | AF339750.1 | - | 1 | 162 | ||
AK295343.1 | - | 2 | 359 | |||
AK313881.1 | - | 2 | 359 | |||
BC007997.1 | - | 2 | 312 | 3357341 | ||
DC331754.1 | - | 2 | 419 |