Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032930.2 | NP_116319.2 | 4 | 402 | ||
XM_005271713.3 | XP_005271770.1 | 4 | 430 | |||
XM_017018454.1 | XP_016873943.1 | 4 | 431 | |||
GenBank mRNA | AK094851.1 | - | 4 | 382 | ||
BC006128.1 | - | 3 | 606 | 3622924 |