Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114172.1 | NP_001107644.1 | 9 | 1241 | ||
NM_001303427.1 | NP_001290356.1 | 8 | 1146 | |||
NM_001303428.1 | NP_001290357.1 | 9 | 1085 | |||
NM_001303429.1 | NP_001290358.1 | 7 | 1563 | |||
NM_003629.3 | NP_003620.3 | 8 | 1740 | |||
XM_017002614.1 | XP_016858103.1 | 9 | 1118 | |||
GenBank mRNA | AF028785.1 | - | 7 | 1498 | ||
AK302049.1 | - | 8 | 1139 | |||
AK304713.1 | - | 3 | 430 | |||
AK313726.1 | - | 9 | 1253 | |||
BC021622.1 | - | 8 | 1674 | 3910221 | ||
CR749649.1 | - | 8 | 1661 | |||
D88532.1 | - | 8 | 1404 |