Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 8 | 973 | ||
NM_152879.2 | NP_690618.2 | 9 | 1038 | |||
XM_005246107.3 | XP_005246164.1 | 7 | 705 | |||
XM_005246108.1 | XP_005246165.1 | 7 | 859 | |||
XM_011512028.2 | XP_011510330.1 | 9 | 2497 | |||
XM_011512029.2 | XP_011510331.1 | 9 | 2498 | |||
XM_011512030.2 | XP_011510332.1 | 9 | 1094 | |||
XM_011512031.2 | XP_011510333.1 | 9 | 968 | |||
XM_011512032.2 | XP_011510334.1 | 9 | 952 | |||
XM_011512033.2 | XP_011510335.1 | 9 | 2499 | |||
XM_011512035.1 | XP_011510337.1 | 7 | 839 | |||
XM_011512037.2 | XP_011510339.1 | 7 | 943 | |||
XM_011512039.2 | XP_011510341.1 | 9 | 2499 | |||
XM_011512040.2 | XP_011510342.1 | 7 | 875 | |||
XM_017005138.1 | XP_016860627.1 | 9 | 1376 | |||
XM_017005139.1 | XP_016860628.1 | 7 | 868 | |||
XM_017005140.1 | XP_016860629.1 | 9 | 970 | |||
GenBank mRNA | AB078966.1 | - | 9 | 1038 | ||
AK308846.1 | - | 7 | 859 | |||
AK311044.1 | - | 7 | 792 | |||
D63479.3 | - | 9 | 979 | |||
D73409.1 | - | 8 | 974 |