Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003655.2 | NP_003646.2 | 4, 5 | 407 | ||
XM_011525399.2 | XP_011523701.1 | 2, 3 | 1916 | |||
GenBank mRNA | AF013956.1 | - | 4, 5 | 410 | ||
AY390430.1 | - | 4, 5 | 244 | |||
BC014967.1 | - | 4, 5 | 263 | 4856728 | ||
BG029608.1 | - | 4, 5 | 240 | |||
BQ225831.1 | - | 407 | ||||
EU439707.1 | - | 4, 5 | 439 | |||
KU178422.1 | - | 4, 5 | 244 | |||
KU178423.1 | - | 4, 5 | 244 |