Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033118.3 | NP_149109.1 | 8 | 1206 | ||
GenBank mRNA | AF325549.1 | - | 7 | 1104 | ||
AJ272502.1 | - | 8 | 1212 | |||
BC007753.1 | - | 1 | 42 | |||
BC069627.1 | - | 7 | 1104 | 7262446 | ||
BC092413.1 | - | 8 | 1206 | 30530456 | ||
BC127622.1 | - | 7 | 1104 | 40085085 | ||
BX647635.1 | - | 8 | 1675 |