Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 5 | 482 | ||
GenBank mRNA | AF268032.1 | - | 5 | 443 | ||
AF423421.1 | - | 5 | 416 | |||
AJ347750.1 | - | 5 | 416 | |||
AK098246.1 | - | 5 | 452 | |||
AK126506.1 | - | 3 | 875 | |||
AK300775.1 | - | 4 | 376 | |||
AK315105.1 | - | 5 | 452 | |||
AL831950.1 | - | 4 | 330 | |||
BC036447.2 | - | 5 | 452 | 4830913 | ||
DC348657.1 | - | 5 | 483 |