Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271999.1 | NP_001258928.1 | 47 | 6236 | ||
NM_001272000.1 | NP_001258929.1 | 47 | 6149 | |||
NM_001272001.1 | NP_001258930.1 | 46 | 6143 | |||
NM_033407.3 | NP_212132.2 | 47 | 6176 | |||
XM_005271292.2 | XP_005271349.1 | 48 | 6232 | |||
XM_011542326.2 | XP_011540628.1 | 48 | 6259 | |||
XM_011542327.2 | XP_011540629.1 | 47 | 6253 | |||
XM_011542328.2 | XP_011540630.1 | 47 | 6244 | |||
XM_017002639.1 | XP_016858128.1 | 46 | 6160 | |||
GenBank mRNA | AB051558.1 | - | 28 | 3652 | ||
AK055493.1 | - | 14 | 1793 | |||
AK055905.1 | - | 19 | 2668 | |||
AK091479.1 | - | 21 | 2643 | |||
AK125049.1 | - | 33 | 4321 | |||
AK299737.1 | - | 19 | 2359 | |||
BC016392.1 | - | 16 | 2140 | |||
BC112387.1 | - | 7 | 895 | |||
DQ118679.1 | - | 44 | 5721 | |||
DQ118680.1 | - | 43 | 5715 | |||
DQ341187.1 | - | 47 | 6143 |