Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 12, 13 | 2167 | ||
XM_005251318.3 | XP_005251375.1 | 11, 12 | 1956 | |||
XM_005251319.4 | XP_005251376.1 | 10, 11 | 1650 | |||
XM_005251320.3 | XP_005251377.1 | 9, 10 | 1442 | |||
XM_005251322.3 | XP_005251379.1 | 8, 9 | 1256 | |||
XM_017013920.1 | XP_016869409.1 | 13, 14 | 2115 | |||
XM_017013923.1 | XP_016869412.1 | 12, 13 | 1942 | |||
XM_017013925.1 | XP_016869414.1 | 10, 11 | 1661 | |||
XM_017013926.1 | XP_016869415.1 | 10, 11 | 1644 | |||
XM_017013927.1 | XP_016869416.1 | 11, 12 | 1672 | |||
XM_017013928.1 | XP_016869417.1 | 8, 9 | 1124 | |||
GenBank mRNA | AB051551.1 | - | 11, 12 | 2788 | ||
AK056185.1 | - | 11, 12 | 2130 | |||
AK057990.1 | - | 3, 4 | 422 | |||
AK300234.1 | - | 9, 10 | 1399 | |||
AK300874.1 | - | 11, 12 | 1811 | |||
AK302648.1 | - | 12, 13 | 2017 | |||
AK307443.1 | - | 9, 10 | 1423 | |||
AK307636.1 | - | 11, 12 | 1801 | |||
BC030701.2 | - | 12, 13 | 2040 | 4837403 |